Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EEF1A1 Recombinant Superclonal™ Antibody

Catalog No. 711669
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-166-9 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-166-9 Supplier Invitrogen™ Supplier No. 711669
Add to Cart
Add to Cart

Rabbit Recombinant Superclonal Antibody

This antibody is predicted to react with Mouse,rat,monkey, Bovine Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called Recombinant polyclonal antibody, this product is now rebranded as Recombinant Superclonal™ antibody. The physical product and the performance remain unchanged.

This protein promotes the GTP-dependent binding of aminoacyl-tRNA to the A-site of ribosomes during protein biosynthesis. With PARP1 and TXK, forms a complex that acts as a T helper 1 (Th1) cell-specific transcription factor and binds the promoter of IFN-gamma to directly regulate its transcription, and is thus involved importantly in Th1 cytokine production.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EEF1A1
Applications Western Blot
Classification Recombinant Superclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Eef1a1
Gene Accession No. P10126, P68104
Gene Alias CCS3; CCS-3; cervical cancer suppressor 3; CTCL tumor antigen; EE1A1; EEF-1; EEF1A; eef1a1; eEF1A-1; eef1a1.S; Eef1a2; Eef1a2l1; eef1as; eef1a-s; EF-1 alpha; EF-1 alpha2; EF1A; EF1A1; EF1a-like protein; EF1alpha; EF-1-alpha-1; EF-1-alpha-S; EF-Tu; elongation factor 1 A-1; elongation factor 1 alpha subunit; elongation factor 1-alpha 1; Elongation factor 1-alpha, somatic form; elongation factor Tu; elongation factor-1 alpha; eukaryotic elongation factor 1 A-1; eukaryotic translation elongation factor 1 alpha 1; eukaryotic translation elongation factor 1 alpha 1 S homeolog; eukaryotic translation elongation factor 1 alpha 1-like 14; eukaryotic translation elongation factor 1 alpha 2 like 1; eukaryotic translation elongation factor 1 alpha, somatic form; glucocorticoid receptor AF-1 specific elongation factor; GRAF 1EF; GRAF-1EF; HNGC:16303; LENG7; leukocyte receptor cluster (LRC) member 7; leukocyte receptor cluster member 7; prostate tumor-inducing protein 1; PTI1; SI; translation elongation factor 1 alpha 1-like 14; XELAEV_18029462mg
Gene Symbols Eef1a1
Host Species Rabbit
Immunogen Peptide corresponding to human EEF1A1 [ 1) 156-172, 2) 322-341].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13627, 1915
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.