Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EEF1A1 Recombinant Rabbit Monoclonal Antibody (JB44-13)
GREENER_CHOICE

Catalog No. PIMA534709
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534709 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534709 Supplier Invitrogen™ Supplier No. MA534709
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: Rat brain tissue, mouse skeletal muscle tissue lysate, rat skin tissue lysate, Daudi, mouse cerebellum tissue lysate, HepG2, HUVEC, SH-SY-5Y, rat skeletal muscle tissue, human tonsil tissue, human liver cancer tissue, human kidney tissue, mouse smooth muscle tissue, THP-1.

This protein promotes the GTP-dependent binding of aminoacyl-tRNA to the A-site of ribosomes during protein biosynthesis. With PARP1 and TXK, forms a complex that acts as a T helper 1 (Th1) cell-specific transcription factor and binds the promoter of IFN-gamma to directly regulate its transcription, and is thus involved importantly in Th1 cytokine production.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EEF1A1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JB44-13
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Eef1a1
Gene Accession No. P68104
Gene Alias CCS3; CCS-3; cervical cancer suppressor 3; CTCL tumor antigen; EE1A1; EEF-1; EEF1A; eef1a1; eEF1A-1; eef1a1.S; Eef1a2; Eef1a2l1; eef1as; eef1a-s; EF-1 alpha; EF-1 alpha2; EF1A; EF1A1; EF1a-like protein; EF1alpha; EF-1-alpha-1; EF-1-alpha-S; EF-Tu; elongation factor 1 A-1; elongation factor 1 alpha subunit; elongation factor 1-alpha 1; Elongation factor 1-alpha, somatic form; elongation factor Tu; elongation factor-1 alpha; eukaryotic elongation factor 1 A-1; eukaryotic translation elongation factor 1 alpha 1; eukaryotic translation elongation factor 1 alpha 1 S homeolog; eukaryotic translation elongation factor 1 alpha 1-like 14; eukaryotic translation elongation factor 1 alpha 2 like 1; eukaryotic translation elongation factor 1 alpha, somatic form; glucocorticoid receptor AF-1 specific elongation factor; GRAF 1EF; GRAF-1EF; HNGC:16303; LENG7; leukocyte receptor cluster (LRC) member 7; leukocyte receptor cluster member 7; prostate tumor-inducing protein 1; PTI1; SI; translation elongation factor 1 alpha 1-like 14; XELAEV_18029462mg
Gene Symbols Eef1a1
Host Species Rabbit
Immunogen Synthetic peptide within Human eEF1A1 aa 413-462.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1915
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.