Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EEF1A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597690
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597690 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597690 Supplier Invitrogen™ Supplier No. PA597690
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EEF1A2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene encodes an isoform of the alpha subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This isoform (alpha 2) is expressed in brain, heart and skeletal muscle, and the other isoform (alpha 1) is expressed in brain, placenta, lung, liver, kidney, and pancreas. This gene may be critical in the development of ovarian cancer. [provided by RefSeq, Mar 2014].
TRUSTED_SUSTAINABILITY

Specifications

Antigen EEF1A2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.47 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EEF1A2
Gene Accession No. Q05639
Gene Alias Eef1a; EEF1A2; eEF1A-2; EEF1AL; EF1A; EF-1-alpha-2; EIEE33; elongation factor 1 A-2; elongation factor 1-alpha 2; Eukaryotic elongation factor 1 A-2; eukaryotic translation elongation factor 1 alpha 2; HS1; MRD38; Ps10; RATPS10; S1; statin S1; Statin-like protein; statin-S1; STN; Stnl; wasted; wst
Gene Symbols EEF1A2
Host Species Rabbit
Immunogen Recombinant Human Elongation factor 1-alpha 2 protein (184-463AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1917
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.