Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EEF1D Monoclonal Antibody (OTI6B3)
GREENER_CHOICE

Catalog No. PIMA527513
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527513 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527513 Supplier Invitrogen™ Supplier No. MA527513
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

EEF1D Monoclonal Antibody for IHC (P), Western Blot

This gene encodes a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This subunit functions as guanine nucleotide exchange factor. It is reported that this subunit interacts with HIV-1 Tat, and thus it represses the translation of host-cell, but not HIV-1, mRNAs. Several alternatively spliced transcript variants encoding multiple isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EEF1D
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6B3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 1% BSA and 0.02% sodium azide; pH 7.3
Gene EEF1D
Gene Accession No. P29692
Gene Alias antigen NY-CO-4; EEF 1D; Eef1d; EF 1 delta; EF 1D; EF1D; EF-1D; EF-1-delta; Elongation factor 1-delta; eukaryotic translation elongation factor 1 delta; eukaryotic translation elongation factor 1 delta (guanine nucleotide exchange protein); FP1047; guanine nucleotide exchange protein; translation elongation factor 1-delta subunit
Gene Symbols EEF1D
Host Species Mouse
Immunogen Full length human recombinant protein of EEF1D produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1936
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.