Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EEF1D Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111323
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111323 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111323 Supplier Invitrogen™ Supplier No. PA5111323
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PRKSQDSRKP LQKKRKRSPK SGLGPADLAL LGLSAERVWL DKSLFDQAES SYRQKLADVA AQAAWPPALA PWGLCTHGNQ VACHHVTWGI WVNKSS Highest antigen sequence indentity to the following orthologs: Mouse - 80%, Rat - 81%.

This gene encodes a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This subunit functions as guanine nucleotide exchange factor. It is reported that this subunit interacts with HIV-1 Tat, and thus it represses the translation of host-cell, but not HIV-1, mRNAs. Several alternatively spliced transcript variants encoding multiple isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EEF1D
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EEF1D
Gene Accession No. P29692
Gene Alias antigen NY-CO-4; EEF 1D; Eef1d; EF 1 delta; EF 1D; EF1D; EF-1D; EF-1-delta; Elongation factor 1-delta; eukaryotic translation elongation factor 1 delta; eukaryotic translation elongation factor 1 delta (guanine nucleotide exchange protein); FP1047; guanine nucleotide exchange protein; translation elongation factor 1-delta subunit
Gene Symbols EEF1D
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human EEF1D. Recombinant protein control fragment (Product #RP-104916).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1936
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.