Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EEF1D Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576656
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This subunit functions as guanine nucleotide exchange factor. It is reported that this subunit interacts with HIV-1 Tat, and thus it represses the translation of host-cell, but not HIV-1, mRNAs. Several alternatively spliced transcript variants encoding multiple isoforms have been found for this gene.
Specifications
EEF1D | |
Polyclonal | |
Unconjugated | |
EEF1D | |
antigen NY-CO-4; EEF 1D; Eef1d; EF 1 delta; EF 1D; EF1D; EF-1D; EF-1-delta; Elongation factor 1-delta; eukaryotic translation elongation factor 1 delta; eukaryotic translation elongation factor 1 delta (guanine nucleotide exchange protein); FP1047; guanine nucleotide exchange protein; translation elongation factor 1-delta subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1936, 300033, 66656 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P29692, P57776, Q68FR9 | |
EEF1D | |
Recombinant full length Human EEF1D. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction