Learn More
Invitrogen™ EEF1E1 Monoclonal Antibody (1494CT639.147.31)

Description
EEF1E1 Monoclonal Antibody for Western Blot, ICC/IF, Flow
Specifications
Specifications
| Antigen | EEF1E1 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1494CT639.147.31 |
| Concentration | 0.7 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | EEF1E1 |
| Gene Accession No. | O43324, Q9D1M4 |
| Gene Alias | 1110003A02Rik; AIMP3; Aminoacyl tRNA synthetase complex-interacting multifunctional protein 3; ARS-interacting multifunctional protein 3; Eef1e1; Elongation factor p18; eukaryotic translation elongation factor 1 epsilon 1; eukaryotic translation elongation factor 1 epsilon-1; Multisynthase complex auxiliary component p18; multisynthetase complex auxiliary component p18; P18; p18 component of aminoacyl-tRNA synthetase complex |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.