Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EEF1E1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590407
Description
Immunogen sequence: MAAAAELSLL EKSLGLSKGN KYSAQGERQI PVLQTNNGPS LTGLTTIAAH LVKQANKEYL LGSTAEEKAI VQQWLEYRVT QVDGHSSKND IHTLLKDLNS; Positive Samples: 293T, HeLa, Mouse brain, Rat brain; Cellular Location: Cytoplasm, Nucleus.
This gene encodes a multifunctional protein that localizes both in the cytoplasm and in the nucleus. In the cytoplasm, the encoded protein is an auxiliary component of the macromolecular aminoacyl-tRNA synthase complex. However, its mouse homolog has been shown to translocate to the nucleus in response to DNA damage and plays a positive role in ATM/ATR-mediated p53 activation. Alternative splicing results in multiple transcript variants.
Specifications
EEF1E1 | |
Polyclonal | |
Unconjugated | |
EEF1E1 | |
1110003A02Rik; AIMP3; Aminoacyl tRNA synthetase complex-interacting multifunctional protein 3; ARS-interacting multifunctional protein 3; Eef1e1; Elongation factor p18; eukaryotic translation elongation factor 1 epsilon 1; eukaryotic translation elongation factor 1 epsilon-1; Multisynthase complex auxiliary component p18; multisynthetase complex auxiliary component p18; P18; p18 component of aminoacyl-tRNA synthetase complex | |
Rabbit | |
Affinity chromatography | |
RUO | |
291057, 66143, 9521 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.36 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
O43324, Q9D1M4 | |
EEF1E1 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human EEF1E1 (NP_0042711). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction