Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EEF1G Antibody (SR1831), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | EEF1G |
---|---|
Clone | SR1831 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
EEF1G Monoclonal antibody specifically detects EEF1G in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
EEF1G | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
eEF-1B gamma, EF-1-gamma, EF1Gelongation factor 1-gamma, eukaryotic translation elongation factor 1 gamma, GIG35, pancreatic tumor-related protein, PRO1608, translation elongation factor eEF-1 gamma chain | |
A synthesized peptide derived from human EEF1G (Uniprot #: P26641) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1831 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
1937 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title