Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EEF2 Monoclonal Antibody (5B6)
GREENER_CHOICE

Catalog No. PIMA515833
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515833 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515833 Supplier Invitrogen™ Supplier No. MA515833
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15833 targets EEF2 in indirect ELISA, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15833 immunogen is purified recombinant fragment of human EEF2 expressed in E. Coli. . MA5-15833 detects EEF2 which has a predicted molecular weight of approximately 95kDa.

Catalyzes the GTP-dependent ribosomal translocation step during translation elongation. During this step, the ribosome changes from the pre-translocational (PRE) to the post-translocational (POST) state as the newly formed A-site-bound peptidyl-tRNA and P-site-bound deacylated tRNA move to the P and E sites, respectively. Catalyzes the coordinated movement of the two tRNA molecules, the mRNA and conformational changes in the ribosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EEF2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 5B6
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene EEF2
Gene Accession No. P13639, P58252
Gene Alias EEF2; EEF-2; Ef2; EF-2; Elongation factor 2; elongation factor-2; eukaryotic translation elongation factor 2; I79_013848; polypeptidyl-tRNA translocase; SCA26; zef2
Gene Symbols EEF2
Host Species Mouse
Immunogen Purified recombinant fragment of human EEF2 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13629, 1938
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.