Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EEF2K Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10087019AF350
Description
EEF2K Polyclonal antibody specifically detects EEF2K in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
EEF2K | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Calcium/calmodulin-dependent eukaryotic elongation factor 2 kinase, calcium/calmodulin-dependent eukaryotic elongation factor-2 kinase, EC 2.7.11, EC 2.7.11.20, eEF-2 kinase, eEF-2Kcalmodulin-dependent protein kinase III, elongation factor-2 kinase, eukaroytic elongation factor 2 kinase, eukaryotic elongation factor 2 kinase, eukaryotic elongation factor-2 kinase, HSU93850, MGC45041 | |
The immunogen recognized by this antibody maps to a region between residue 675 and 725 of human calcium/calmodulin-dependent eukaryotic elongation factor-2 kinase (EEF2K ) using the numbering given in entry NP_037434.1 (GeneID 29904). | |
0.1 mL | |
Cancer, Hypoxia Signaling, Phospho Specific | |
29904 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction