Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EEF2K Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323302
Description
EEF2K Polyclonal antibody specifically detects EEF2K in Human, Mouse, Rat, Chicken, Xenopus samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
EEF2K | |
Polyclonal | |
Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:100 | |
Calcium/calmodulin-dependent eukaryotic elongation factor 2 kinase, calcium/calmodulin-dependent eukaryotic elongation factor-2 kinase, EC 2.7.11, EC 2.7.11.20, eEF-2 kinase, eEF-2Kcalmodulin-dependent protein kinase III, elongation factor-2 kinase, eukaroytic elongation factor 2 kinase, eukaryotic elongation factor 2 kinase, eukaryotic elongation factor-2 kinase, HSU93850, MGC45041 | |
eEF2K (C-terminus) synthetic peptide (coupled to carrier protein) corresponds to amino acids in the C-terminal region of human eEF2K. This sequence is well conserved in rat, mouse, chicken, and fish eEF2K, and has low homology to other proteins. [UniProt# O00418] | |
0.1 mL | |
Cancer, Hypoxia Signaling, Phospho Specific | |
29904 | |
Store at -20°C. | |
IgG |
Western Blot, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Chicken, Xenopus species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction