Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EEF2K Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501728797 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-8797 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-8797 Supplier Proteintech Group Inc Supplier No. 135101AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CD54, M.W. 90,000, antigen is the Intercellular Adhesion Molecule-1 (ICAM-1) and is the ligand for LFA-1 mediated adhesion. CD54 is weakly expressed on peripheral blood leukocytes. Upon activation the antigen is expressed strongly on B cells, T cells, macrophages and granulocytes. ICAM-1 is also inducible on endothelial cells, epithelial cells and fibroblasts.

Specifications

Antigen EEF2K
Applications Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EEF2K
Gene Accession No. O00418, O08796, P70531
Gene Alias eEF 2 kinase, eEF 2K, EEF2K, HSU93850
Gene Symbols Eef2k
Host Species Rabbit
Immunogen EEF2K Fusion Protein Ag4432
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13631, 25435, 29904
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.