Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EEFSEC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557400
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557400 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557400 Supplier Invitrogen™ Supplier No. PA557400
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AKFHITVGHE TVMGRLMFFS PAPDNFDQEP ILDSFNFSQE YLFQEQYLSK DLTPAVTDND EADKKAGQAT EGHCPRQQWA LVEFEKPVTC Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 80%.

SELB is a translation factor necessary for the incorporation of selenocysteine into proteins. It probably replaces EF-Tu for the insertion of selenocysteine directed by the UGA codon. SelB binds GTP and GDP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EEFSEC
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EEFSEC
Gene Accession No. P57772
Gene Alias Eefsec; EFSEC; elongation factor for selenoprotein translation; elongation factor sec; eukaryotic elongation factor, selenocysteine-tRNA specific; eukaryotic elongation factor, selenocysteine-tRNA-specific; MJ0495-like protein SelB; mSelB; RGD1560552; sec; SELB; selenocysteine (Sec)-specific eukaryotic elongation factor; selenocysteine-specific elongation factor
Gene Symbols EEFSEC
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EEFSEC. Recombinant protein control fragment (Product #RP-96337).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60678
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.