Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EEPD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565812
Description
Immunogen sequence: SVEDLVRMDGI NAAFLDRIRH QVFAERSRPP STHTNGGLTF TAKPHPSPTS LSLQSEDLDL PPGGPTQIIS TRPSVEAFGG TRDGRPVLRL ATWNLQGCSV Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.
EEPD1 gene ontology annotations related to this gene include DNA repair.
Specifications
EEPD1 | |
Polyclonal | |
Unconjugated | |
EEPD1 | |
2310005P05Rik; AI481005; EEPD1; endonuclease/exonuclease/phosphatase family domain containing 1; endonuclease/exonuclease/phosphatase family domain-containing protein 1; HSPC107; hypothetical protein LOC511767; KIAA1706; KIAA1706 protein; RGD1307401 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80820 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7L9B9 | |
EEPD1 | |
Recombinant Human EEPD1. Recombinant protein control fragment (Product #RP-104973). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction