Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EEPD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569536
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569536 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569536 Supplier Invitrogen™ Supplier No. PA569536
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

EEPD1 gene ontology annotations related to this gene include DNA repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EEPD1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene EEPD1
Gene Accession No. Q3TGW2
Gene Alias 2310005P05Rik; AI481005; EEPD1; endonuclease/exonuclease/phosphatase family domain containing 1; endonuclease/exonuclease/phosphatase family domain-containing protein 1; HSPC107; hypothetical protein LOC511767; KIAA1706; KIAA1706 protein; RGD1307401
Gene Symbols EEPD1
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human EEPD1 (aa 1-50).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67484
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.