Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EEPD1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS14506R

Description
EEPD1 gene ontology annotations related to this gene include DNA repair.Specifications
EEPD1 | |
Polyclonal | |
Unconjugated | |
EEPD1 | |
2310005P05Rik; AI481005; BOS_4284; EEPD1; endonuclease/exonuclease/phosphatase family domain containing 1; Endonuclease/exonuclease/phosphatase family domain-containing protein 1; HSPC107; KIAA1706; RGD1307401 | |
Rabbit | |
Protein A | |
RUO | |
80820 | |
-20°C | |
Liquid |
Western Blot | |
1 μg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH | |
Q7L9B9 | |
EEPD1 | |
KLH conjugated synthetic peptide derived from human EEPD1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction