Learn More
EF-1 delta Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301684A
Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This subunit functions as guanine nucleotide exchange factor. It is reported that this subunit interacts with HIV-1 Tat, and thus it represses the translation of host-cell, but not HIV-1, mRNAs. Several alternatively spliced transcript variants encoding multiple isoforms have been found for this gene.Specifications
| EF-1 delta | |
| Polyclonal | |
| Unconjugated | |
| EEF1D | |
| Elongation factor 1-delta; EF-1-delta; Antigen NY-CO-4; EF1D; EEF1D | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1936 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| P29692 | |
| EEF1D | |
| Between 80 and 130 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.