Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EFCAB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554813
Description
Immunogen sequence: YCFEVFDLNG DGFISKEEMF HMLKNSLLKQ PSEEDPDEGI KDLVEITLKK MDHDHDGKLS FADYELAVRE ETLLLEAFGP CLPDPKSQME FEAQVFKDPN Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
EFCAB1 gene ontology annotations related to this gene include calcium ion binding.
Specifications
EFCAB1 | |
Polyclonal | |
Unconjugated | |
EFCAB1 | |
EFCAB1; EF-hand calcium binding domain 1; EF-hand calcium-binding domain-containing protein 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79645 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HAE3 | |
EFCAB1 | |
Recombinant protein corresponding to Human EFCAB1. Recombinant protein control fragment (Product #RP-93728). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction