Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EFCAB11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565652
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565652 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565652 Supplier Invitrogen™ Supplier No. PA565652
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MFFSEARARSR TWEASPSEHR KWVEVFKACD EDHKGYLSRE DFKTAVVMLF GYKPSKIEVD SVMSS Highest antigen sequence identity to the following orthologs - mouse 80%, rat 80%.

EFCAB11 gene ontology annotations related to this gene include calcium-mediated signaling; microtubule nucleation; mitotic spindle pole body duplication; spindle pole body duplication.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EFCAB11
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EFCAB11
Gene Accession No. Q9BUY7
Gene Alias C14orf143; EFCAB11; EF-hand calcium binding domain 11; EF-hand calcium-binding domain-containing protein 11; EF-hand domain-containing protein C14orf143
Gene Symbols EFCAB11
Host Species Rabbit
Immunogen Recombinant Human EFCAB11. Recombinant protein control fragment (Product #RP-106328).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90141
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.