Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EFCAB12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5139835
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: DKVCPIRQPG GYYSDWKVFS PNLALLRSQG PGKSKRTDKK TPKKSKKMRF KEFEEFTRKL KVKRSSGLQQ THPNSFWPGH L.
EFCAB12 is a protein coding gene.
Specifications
| EFCAB12 | |
| Polyclonal | |
| Unconjugated | |
| EFCAB12 | |
| C3orf25; EFCAB12; EF-hand calcium binding domain 12; EF-hand calcium-binding domain-containing protein 12; EF-hand domain-containing protein C3orf25 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90288 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6NXP0 | |
| EFCAB12 | |
| Recombinant protein corresponding to Human EFCAB12. Recombinant protein control fragment (Product #RP-108943). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction