Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EFCAB12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558064
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558064 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558064 Supplier Invitrogen™ Supplier No. PA558064
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NPASQPQAPP KPIPSFKVLE ARDIQEQPED RKTWLSQRSK LRQELESFGD VKRWLENKPS ITPSEAKVLH MIHEEQSAQP NASQATTRTT Highest antigen sequence identity to the following orthologs: Mouse - 49%, Rat - 52%.

EFCAB12 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EFCAB12
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EFCAB12
Gene Accession No. Q6NXP0
Gene Alias C3orf25; EFCAB12; EF-hand calcium binding domain 12; EF-hand calcium-binding domain-containing protein 12; EF-hand domain-containing protein C3orf25
Gene Symbols EFCAB12
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EFCAB12. Recombinant protein control fragment (Product #RP-96457).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90288
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.