Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EFCAB13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554476
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554476 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554476 Supplier Thermo Scientific Supplier No. PA554476
Add to Cart
Add to Cart

EFCAB13 Polyclonal Antibody

EFCAB13 Polyclonal Antibody for ICC/IF, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen EFCAB13
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EFCAB13
Gene Accession No. Q8IY85
Gene Alias C17orf57; EFCAB13; EF-hand calcium binding domain 13; EF-hand calcium-binding domain-containing protein 13; EF-hand domain-containing protein C17orf57
Gene Symbols EFCAB13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EFCAB13. Recombinant protein control fragment (Product #RP-93468).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124989
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.