Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EFCAB14 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$487.50

Specifications

Antigen KIAA0494
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP159967
SDP
View Documents
Novus Biologicals
NBP159967
100 μL
Each of 1 for $487.50
Only null left
Add to Cart
 
Description

Description

EFCAB14 Polyclonal specifically detects EFCAB14 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

KIAA0494
Polyclonal
Purified
RUO
hypothetical protein LOC9813, KIAA0494, RP11-8J9.3
EFCAB14
IgG
Protein A purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
O75071
9813
Synthetic peptides corresponding to KIAA0494(KIAA0494) The peptide sequence was selected from the N terminal of KIAA0494. Peptide sequence DLDALKEKFRTMESNQKSSFQEIPKLNEELLSKQKQLEKIESGEMGLNKV.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.