Learn More
Invitrogen™ EFCAB14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552807
Description
Immunogen sequence: ESDVVAMSKV EKKANLSFSM MGDRSATLKR QSLDQVTNRT DTVKIQSIKK EDSSNSQVSK LREKLQLISA LTNKPESNRP PETADEEQVE SFTSKPSALP KFSQFLGDPV EKAAQLRPIS LPGVSSTEDL QDLFRKTGQD VDGKLTYQE Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 77%.
EFCAB14 gene ontology annotations related to this gene include calcium ion binding.
Specifications
EFCAB14 | |
Polyclonal | |
Unconjugated | |
EFCAB14 | |
4732418C07Rik; EFCAB14; EF-hand calcium binding domain 14; EF-hand calcium-binding domain-containing protein 14; Kiaa0494; RGD1310351; RP11-8J9.3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9813 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75071 | |
EFCAB14 | |
Recombinant protein corresponding to Human EFCAB14. Recombinant protein control fragment (Product #RP-90357). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.