Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EFCAB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556673
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556673 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556673 Supplier Invitrogen™ Supplier No. PA556673
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SLGISQATGS AARWRTRRTG KGLGYNSDEI RPRTLLIEHL MEGGRRDHHT MTVLWGTQEI IVAEFHKKIK EAFEVFDHES NNTVDVR Highest antigen sequence identity to the following orthologs: Mouse - 28%, Rat - 31%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen EFCAB2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EFCAB2
Gene Accession No. Q5VUJ9
Gene Alias CFAP200; DRC8; Dynein regulatory complex protein 8; dynein regulatory complex subunit 8; EFCAB2; EF-hand calcium binding domain 2; EF-hand calcium-binding domain-containing protein 2
Gene Symbols EFCAB2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EFCAB2. Recombinant protein control fragment (Product #RP-94559).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84288
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.