Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EFCAB4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561187
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561187 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561187 Supplier Invitrogen™ Supplier No. PA561187
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEAVFESLDQ AHTGFLTARE FCLGLGMFVG VASAQGANPC RTPEETFESG GLDVQGTAGS LDEE Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 77%.

EFCAB4A, also known as Calcium release-activated calcium channel regulator 2B, is a novel Ca2+-binding EF-hand protein that is thought to play a key role in store-operated Ca2+ entry in T-cells by regulating CRAC channel activation, but the detailed function is still under investigation. It is likely to play a similar role as the related protein EFCAB4B, which acts as a cytoplasmic calcium-sensor that forms a complex with ORAI1 and STIM1 at the junctional regions between the plasma membrane and the endoplasmic reticulum upon low Ca2+ concentration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EFCAB4A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CRACR2B
Gene Accession No. Q8N4Y2
Gene Alias 6330520A15Rik; BC026645; Ca2+ release-activated Ca2+ (CRAC) channel regulator 2B; calcium release activated channel regulator 2B; calcium release-activated calcium channel regulator 2B; Calcium release-activated channel regulator 2B; CRAC channel regulator 2B; CRAC regulator 2B; Cracr2a; CRACR2B; Efcab4; EFCAB4A; EF-hand calcium binding domain 4A; EF-hand calcium-binding domain-containing protein 4A; RGD1560911
Gene Symbols CRACR2B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EFCAB4A. Recombinant protein control fragment (Product #RP-97506).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283229
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.