Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EFCAB4B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558493
Description
Immunogen sequence: PDGRVVSRPQ RLGQGSGQGP KGSGACLHPL DSLEQKETQE QTSGQLVMLR KAQEFFQTCD AEGKGFIARK DMQRLHKELP LSLEELEDVF DALDA Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 59%.
CRACR2A is a Ca(2+)-binding protein that plays a key role in store-operated Ca(2+) entry (SOCE) in T-cells by regulating CRAC channel activation. CRACR2A acts as a cytoplasmic calcium-sensor that facilitates the clustering of ORAI1 and STIM1 at the junctional regions between the plasma membrane and the endoplasmic reticulum upon low Ca(2+) concentration. Further, CRACR2A regulates CRAC channel activation, including translocation and clustering of ORAI1 and STIM1. Upon increase of cytoplasmic Ca(2+) resulting from opening of CRAC channels, CRACR2A dissociates from ORAI1 and STIM1, thereby destabilizing the ORAI1-STIM1 complex.
Specifications
EFCAB4B | |
Polyclonal | |
Unconjugated | |
Cracr2a | |
Ca2+ release-activated Ca2+ (CRAC) channel regulator 2A; calcium release activated channel regulator 2A; calcium release-activated calcium channel regulator 2A; Calcium release-activated channel regulator 2A; CRAC channel regulator 2A; CRAC regulator 2A; Cracr2a; Efcab4b; EF-hand calcium binding domain 4B; EF-hand calcium-binding domain-containing protein 4B; Gm1073; Gm462; RGD1563719 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84766 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BSW2 | |
Cracr2a | |
Recombinant protein corresponding to Human EFCAB4B. Recombinant protein control fragment (Product #RP-97745). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction