Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EFCAB4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589473
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589473 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589473 Supplier Invitrogen™ Supplier No. PA589473
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAAPDGRVVS RPQRLGQGSG QGPKGSGACL HPLDSLEQKE TQEQTSGQLV MLRKAQEFFQ TCDAEGKGFI ARKDMQRLHK ELPLSLEELE DVFDALDADG NGYLTPQEFT TGFSHFFFSQ NNPSQEDAGE QVAQRHEEKV YLSRGDEDLG DMGEDEEAQF RMLMDRLGAQ KVLEDESDVK QLWLQLKKEE PHLLSNFEDF; Positive Samples: Mouse liver; Cellular Location: Cytoplasm.

CRACR2A is a Ca(2+)-binding protein that plays a key role in store-operated Ca(2+) entry (SOCE) in T-cells by regulating CRAC channel activation. CRACR2A acts as a cytoplasmic calcium-sensor that facilitates the clustering of ORAI1 and STIM1 at the junctional regions between the plasma membrane and the endoplasmic reticulum upon low Ca(2+) concentration. Further, CRACR2A regulates CRAC channel activation, including translocation and clustering of ORAI1 and STIM1. Upon increase of cytoplasmic Ca(2+) resulting from opening of CRAC channels, CRACR2A dissociates from ORAI1 and STIM1, thereby destabilizing the ORAI1-STIM1 complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EFCAB4B
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.01 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Cracr2a
Gene Accession No. Q3UP38, Q9BSW2
Gene Alias Ca2+ release-activated Ca2+ (CRAC) channel regulator 2A; calcium release activated channel regulator 2A; calcium release-activated calcium channel regulator 2A; Calcium release-activated channel regulator 2A; CRAC channel regulator 2A; CRAC regulator 2A; Cracr2a; Efcab4b; EF-hand calcium binding domain 4B; EF-hand calcium-binding domain-containing protein 4B; Gm1073; Gm462; RGD1563719
Gene Symbols Cracr2a
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human CRACR2A (NP_0011384301).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 381812, 84766
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.