Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EFCAB4B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521060
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521060 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521060 Supplier Thermo Scientific Supplier No. PA521060
Add to Cart
Add to Cart

EFCAB4B Polyclonal Antibody

EFCAB4B Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen EFCAB4B
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Cracr2a
Gene Accession No. Q3UP38, Q9BSW2
Gene Alias Ca2+ release-activated Ca2+ (CRAC) channel regulator 2A; calcium release activated channel regulator 2A; calcium release-activated calcium channel regulator 2A; Calcium release-activated channel regulator 2A; CRAC channel regulator 2A; CRAC regulator 2A; Cracr2a; Efcab4b; EF-hand calcium binding domain 4B; EF-hand calcium-binding domain-containing protein 4B; Gm1073; Gm462; RGD1563719
Gene Symbols Cracr2a
Host Species Rabbit
Immunogen A 14 amino acid peptide near the carboxy terminus of human EFCAB4B.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 381812, 84766
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.