Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EFCAB6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546780
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546780 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546780 Supplier Invitrogen™ Supplier No. PA546780
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: FGLKATTKIN WKQFLTSFHE PQGLQVSSKG PLTKRNSINS RNESHKENII.

This gene encodes a protein which directly binds the oncogene DJ-1 and androgen receptor to form a ternary complex in cells. This binding protein recruits histone-deacetylase complexes in order to repress transcription activity of androgen receptor. This protein may also play a role in spermatogenesis and fertilization. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EFCAB6
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene EFCAB6
Gene Accession No. Q5THR3
Gene Alias CAP-binding protein complex-interacting protein 1; dJ185D5.1; DJ-1-binding protein; DJBP; EFCAB6; EF-hand calcium binding domain 6; EF-hand calcium-binding domain-containing protein 6; HSCBCIP1; KIAA1672
Gene Symbols EFCAB6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human EFCAB6 (aa 345-394).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64800
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.