Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EFCAB7 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP216276
Description
EFCAB7 Polyclonal specifically detects EFCAB7 in Human samples. It is validated for Western Blot.Specifications
EFCAB7 | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
DKFZp666D225, EF-hand calcium binding domain 7, EF-hand calcium-binding domain-containing protein 7, KIAA1799RP4-534K7.1 | |
Rabbit | |
72 kDa | |
0.1 mL | |
Primary | |
Human. Expected to cross react based on sequence identity: Mouse (86%), Rat (83%), Bovine (86%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot | |
1mg/mL | |
Western Blot 1:500-1:3000 | |
A8K855 | |
EFCAB7 | |
Recombinant protein encompassing a sequence within the C-terminus region of human EFCAB7. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
84455 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction