Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EFCAB7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597696
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597696 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597696 Supplier Invitrogen™ Supplier No. PA597696
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

EFCAB7 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

EFCAB7 (EF-Hand Calcium Binding Domain 7) is a Protein Coding gene. Diseases associated with EFCAB7 include Ellis-Van Creveld Syndrome. Among its related pathways are Signaling by Hedgehog and Signaling by GPCR. Gene Ontology (GO) annotations related to this gene include calcium ion binding. An important paralog of this gene is CABP2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EFCAB7
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene EFCAB7
Gene Accession No. A8K855, Q8VDY4
Gene Alias EFCAB7; EF-hand calcium binding domain 7; EF-hand calcium-binding domain-containing protein 7; Kiaa1799; mKIAA1799; RP4-534K7.1
Gene Symbols EFCAB7
Host Species Rabbit
Immunogen Recombinant Human EF-hand calcium-binding domain-containing protein 7 protein (1-158AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230500, 84455
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.