Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EFCAB7 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | EFCAB7 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
EFCAB7 Polyclonal specifically detects EFCAB7 in Human, Mouse samples. It is validated for Western Blot.Specifications
EFCAB7 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
84455 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
DKFZp666D225, EF-hand calcium binding domain 7, EF-hand calcium-binding domain-containing protein 7, KIAA1799RP4-534K7.1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 170-400 of human EFCAB7 (NP_115813.2). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title