Learn More
Invitrogen™ EFEMP2 Polyclonal Antibody

Description
Antibody detects endogenous levels of total EFEMP2.
Specifications
Specifications
| Antigen | EFEMP2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Efemp2 |
| Gene Accession No. | O95967 |
| Gene Alias | 0610011K11Rik; ARCL1B; Efemp2; EGF containing fibulin extracellular matrix protein 2; EGF containing fibulin like extracellular matrix protein 2; EGF containing fibulin-like extracellular matrix protein 2; EGF-containing fibulin-like extracellular matrix protein 2; EGF-containing fibulin-like extracellular matrix protein 2 (Efemp2); epidermal growth factor-containing fibulin-like extracellular matrix protein 2; Fbln4; FIBL-4; fibulin 4; Fibulin4; fibulin-4; Mbp1; mutant p53 binding protein 1; mutant p53-binding protein 1; Protein UPH1; UNQ200/PRO226; UPH1 |
| Gene Symbols | Efemp2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.