Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Innovating Science™ Effectiveness of Sunscreen

Shop All Aldon Products


Click to view available options
Class Size:
15 Lab Groups
Description
Includes
- 15 mL each of SPF 15 and SPF 30 sunscreen
- One box of microscope slides (enough for 15 setups)
- Instructions
Requires
NeuLog™ UVA Logger Sensor (S98243-36ND) and UVB Logger Sensor (SS98243-37ND); sensors not included
- Analyze the difference in sunscreens — discover if SPF 15 is really different from SPF 30
- Kit contains enough reagents for 15 setups
Specifications
Specifications
Color | White |
Grade | 7 to 14 |
Age | 13 to 20 |
For Use With (Application) | Study both sunscreens and sun radiation |
Includes | 15 mL each of spf 15 and spf 30 sunscreen, one box of microscope slides (enough for 15 setups), instructions |
Quantity | 1 |
Dimensions (L x W x H) | 9.75 x 6.75 x 3 in. |
Class Size | 15 Lab Groups |
Product Type | Effectiveness of Sunscreen Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction