Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EFHA1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561023
Description
Immunogen sequence: LAAAVAGAAL AGAGAAWHHS RVSVAARDGS FTVSAQKNVE HGIIYIGKPS LRKQRFMQFS SLEHEGEYYM TPRDFLFSVM FEQMERKTSV KKLTKKDIED TLSGIQT Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 76%.
Key regulator of mitochondrial calcium uniporter (MCU) required to limit calcium uptake by MCU when cytoplasmic calcium is low. MICU1 and MICU2 form a disulfide-linked heterodimer that stimulate and inhibit MCU activity, depending on the concentration of calcium. MICU2 acts as a gatekeeper of MCU that senses calcium level via its EF-hand domains: prevents channel opening at resting calcium, avoiding energy dissipation and cell-death triggering.
Specifications
EFHA1 | |
Polyclonal | |
Unconjugated | |
MICU2 | |
09C01; 1110008L20Rik; 4833427E09Rik; AU041871; Calcium uptake protein 2, mitochondrial; EF hand domain family A1; EF hand domain family, member A1; EFHA1; EF-hand domain family member A1; EF-hand domain family, member A1; EF-hand domain-containing family member A1; Micu2; mitochondrial calcium uptake 2; RGD1309934; Smhs2; Smhs2 homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
221154 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IYU8 | |
MICU2 | |
Recombinant protein corresponding to Human EFHA1. Recombinant protein control fragment (Product #RP-97982). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction