Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EFHA1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988749
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8749 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8749 Supplier Bioss Supplier No. BS9011R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Key regulator of mitochondrial calcium uniporter (MCU) required to limit calcium uptake by MCU when cytoplasmic calcium is low. MICU1 and MICU2 form a disulfide-linked heterodimer that stimulate and inhibit MCU activity, depending on the concentration of calcium. MICU2 acts as a gatekeeper of MCU that senses calcium level via its EF-hand domains: prevents channel opening at resting calcium, avoiding energy dissipation and cell-death triggering.

Specifications

Antigen EFHA1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MICU2
Gene Accession No. Q8IYU8
Gene Alias 09C01; 1110008L20Rik; 4833427E09Rik; AU041871; Calcium uptake protein 2, mitochondrial; EF hand domain family A1; EF hand domain family, member A1; Efha1; EF-hand domain family member A1; EF-hand domain family, member A1; EF-hand domain-containing family member A1; MICU2; mitochondrial calcium uptake 2; RGD1309934; Smhs2; Smhs2 homolog
Gene Symbols MICU2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human EFHA1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221154
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.