Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EFHB Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP26905925UL
Description
EFHB Polyclonal antibody specifically detects EFHB in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
EFHB | |
Polyclonal | |
Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
EF-hand domain family, member B, EF-hand domain-containing family member B, FLJ25200 | |
This antibody was developed against a recombinant protein corresponding to amino acids: KVIPVGYRVATCLTEKLPRLITPPEAKKYFNFRYPPAGVERVFYGRANDPQIAPYLTHGIRSKISILANTLINPQPITTFQQKIKDKKESIYLSNRRAPL | |
25 μL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol | |
Rabbit | |
Protein A purified | |
RUO | |
151651 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction