Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EFHB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585049
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585049 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585049 Supplier Invitrogen™ Supplier No. PA585049
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KVIPVGYRVA TCLTEKLPRL ITPPEAKKYF NFRYPPAGVE RVFYGRANDP QIAPYLTHGI RSKISILANT LINPQPITTF QQKIKDKKES IYLSNRRAPL Highest antigen sequence indentity to the following orthologs: Mouse - 89%, Rat - 81%.

EFHB contains two EF-hand domains. There are three named isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EFHB
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EFHB
Gene Accession No. Q8N7U6
Gene Alias CFAP21; cilia and flagella associated protein 21; EF-hand domain family member B; EF-hand domain family, member B; EF-hand domain-containing family member B; EFHB
Gene Symbols EFHB
Host Species Rabbit
Immunogen Recombinant protein fragment of Human EFHB (Product #RP-108699)
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151651
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.