Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EFHC1 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP320220AF350
Description
EFHC1 Polyclonal antibody specifically detects EFHC1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
EFHC1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
dJ304B14.2, EF-hand domain (C-terminal) containing 1, EF-hand domain-containing protein 1, EJA1, EJM1, FLJ10466, FLJ37290, JAE, Myoclonin-1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human EFHC1 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
114327 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction