Learn More
Invitrogen™ EFHC1 Monoclonal Antibody (4E7)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA524511
Catalog No. PIMA524511
Only null left
Add to Cart
Description
Description
Peptide Sequence: DDTVEIREVH ERNDGRDPFP LLMNRQRVPK VLVENAKNFP QCVLEISDQE VLEWYTAKDF IVGKSLTILG RTFFIYDCDP FTRRYYKEKF GITDLPRIDV SKREPPPVK.
This gene encodes an EF-hand-containing calcium binding protein. The encoded protein likely plays a role in calcium homeostasis. Mutations in this gene have been associated with susceptibility to juvenile myoclonic epilepsy and juvenile absence epilepsy. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EFHC1
Monoclonal
Unconjugated
EFHC1
1700029F22Rik; dJ304B14.2; EF-hand domain (C-terminal) containing 1; EF-hand domain containing 1; EF-hand domain-containing protein 1; EFHC1; EJM1; mRib72-1; myoclonin1; myoclonin-1
Mouse
Protein A
RUO
114327
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
4E7
PBS with no preservative; pH 7.4
Q5JVL4
EFHC1
EFHC1 (NP_060570.1, 270 a.a. ~ 378 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2a κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.