Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EFHC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557219
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557219 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557219 Supplier Invitrogen™ Supplier No. PA557219
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ESKQTEKDPG VQELEALIDT IQKQLKDHSC KDNIREAFQI YDKEASGYVD RDMFFKICES LNVPVDDSLV KELIRMCSHG EGKINYYNFV RAF Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 66%.

This gene encodes an EF-hand-containing calcium binding protein. The encoded protein likely plays a role in calcium homeostasis. Mutations in this gene have been associated with susceptibility to juvenile myoclonic epilepsy and juvenile absence epilepsy. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EFHC1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EFHC1
Gene Accession No. Q5JVL4
Gene Alias 1700029F22Rik; dJ304B14.2; EF-hand domain (C-terminal) containing 1; EF-hand domain containing 1; EF-hand domain-containing protein 1; EFHC1; EJM1; mRib72-1; myoclonin1; myoclonin-1
Gene Symbols EFHC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EFHC1. Recombinant protein control fragment (Product #RP-102660).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114327
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.