Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EFHC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596895
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596895 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596895 Supplier Invitrogen™ Supplier No. PA596895
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EDSAQNCFAL IPKAPKKDVI KMLVNDNKVL RYLAVLESPI PEDKDRRFVF SYFLATDMIS IFEPPVRNSG IIGGKYLGRT KVVKPYSTVD NPVYYGPSDF FIGAVIEVFG HRFIILDTDE YVLKYMESNA AQYSPEALAS IQNHVRKREA PAPEAESKQT EKDPGVQELE ALIDTIQKQL KDHSCKDNIR EAFQIYDKEA SGYVDRDMFF KICESLNVPV DDSLVKELIR MCSHGEGKIN YYNFVRAFSN; Positive Samples: HeLa, Mouse testis, Mouse brain; Cellular Location: Cytoplasm, centrosome, cytoskeleton, microtubule organizing center, spindle.

This gene encodes an EF-hand-containing calcium binding protein. The encoded protein likely plays a role in calcium homeostasis. Mutations in this gene have been associated with susceptibility to juvenile myoclonic epilepsy and juvenile absence epilepsy. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EFHC1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene EFHC1
Gene Accession No. Q5JVL4, Q9D9T8
Gene Alias 1700029F22Rik; dJ304B14.2; EF-hand domain (C-terminal) containing 1; EF-hand domain containing 1; EF-hand domain-containing protein 1; EFHC1; EJM1; mRib72-1; myoclonin1; myoclonin-1
Gene Symbols EFHC1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 391-640 of human EFHC1 (NP_0605702).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114327, 301295, 71877
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.