Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EFHD2 Antibody, Novus Biologicals™
SDP

Goat Polyclonal Antibody has been used in 3 publications

$231.00 - $499.50

Specifications

Antigen EFHD2
Concentration 0.5 mg/mL
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB1001477
SDP
View Documents
Novus Biologicals
NB1001477
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
NB407716
SDP
View Documents
Novus Biologicals
NB100147725UG
25 μg
Each for $231.00
Only null left
Add to Cart
 
Description

Description

EFHD2 Polyclonal specifically detects EFHD2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications

Specifications

EFHD2
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Unconjugated
RUO
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
EF hand domain containing 2, EF hand domain family, member D2, EF-hand domain family, member D2, EF-hand domain-containing protein D2, MGC4342, swiprosin 1, swiprosin-1, SWS1
EFHD2
IgG
Affinity Purified
27 kDa
0.5 mg/mL
Polyclonal
Goat
Human
Q96C19
79180
Peptide with sequence CAAFKELQSTFK corresponding to C-Terminus according to NP_077305.2.
Primary
This antibody does not cross-react with Mouse Swiprosin 2. Data kindly provided by Sebastian Duetting and Dirk Mielenz.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.