Learn More
Invitrogen™ EFHD2 Polyclonal Antibody

Description
Immunogen sequence: MATDELATKL SRRLQMEGEG GGETPEQPGL NGAAAAAAGA PDEAAEALGS ADCELSAKLL RRADLNQGIG EPQSPSRR; Positive Samples: Mouse lung; Cellular Location: Membrane raft.
Specifications
Specifications
| Antigen | EFHD2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.48 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | Efhd2 |
| Gene Accession No. | Q9D8Y0 |
| Gene Alias | 2600015J22Rik; AA408606; D4Wsu27e; EF hand domain containing 2; EF hand domain family, member D2; EF-hand domain family member D2; EF-hand domain family, member D2; EF-hand domain-containing protein D2; Efhd2; MGC4342; RP3-467K16.3; swiprosin 1; swiprosin-1; Sws1; testicular tissue protein Li 62 |
| Gene Symbols | Efhd2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.