Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EFR3A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989239
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9239 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9239 Supplier Bioss Supplier No. BS9662R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen EFR3A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene EFR3A
Gene Accession No. Q14156
Gene Alias A130089M23Rik; BB071175; C76891; C920006C10Rik; D030063F01Rik; EFR3 homolog A; EFR3 homolog A (S. cerevisiae); Efr3a; KIAA0143; mKIAA0143; Protein EFR3 homolog A; Protein EFR3-like; RGD1305976
Gene Symbols EFR3A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human EFR3A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23167
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.