Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EFR3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114282
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114282 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114282 Supplier Invitrogen™ Supplier No. PA5114282
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VPSLLFNLQH VEEAESRSPS PLQAPEKEKE SPAELAERCL RELLGRAAFG For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%; Zebrafish: 93%.

Component of a complex required to localize phosphatidylinositol 4-kinase (PI4K) to the plasma membrane. The complex acts as a regulator of phosphatidylinositol 4-phosphate (PtdIns(4)P) synthesis (Probable). In the complex, EFR3B probably acts as the membrane-anchoring component. Also involved in responsiveness to G-protein-coupled receptors; it is however unclear whether this role is direct or indirect.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EFR3B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene EFR3B
Gene Accession No. Q9Y2G0
Gene Alias AI852640; C030014M07Rik; EFR3 homolog B; EFR3 homolog B (S. cerevisiae); Efr3b; FLJ37871; Kiaa0953; mKIAA0953; Protein EFR3 homolog B; RGD1310845
Gene Symbols EFR3B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal region of Human EFR3B (aa 197-246).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22979
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.