Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EFR3B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139908
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139908 Supplier Invitrogen™ Supplier No. PA5139908
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: DRLLSTALME DAEIRLFVLE ILISFIDRHG NRHKFSTIST LSDISVLKLK VDKCSRQDTV FMKKHSQQL Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 99%.

Component of a complex required to localize phosphatidylinositol 4-kinase (PI4K) to the plasma membrane. The complex acts as a regulator of phosphatidylinositol 4-phosphate (PtdIns(4)P) synthesis (Probable). In the complex, EFR3B probably acts as the membrane-anchoring component. Also involved in responsiveness to G-protein-coupled receptors; it is however unclear whether this role is direct or indirect.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EFR3B
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EFR3B
Gene Accession No. Q9Y2G0
Gene Alias AI852640; C030014M07Rik; EFR3 homolog B; EFR3 homolog B (S. cerevisiae); Efr3b; FLJ37871; Kiaa0953; mKIAA0953; Protein EFR3 homolog B; RGD1310845
Gene Symbols EFR3B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human EFR3B. Recombinant protein control fragment (Product #RP-109266).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22979
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.