Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EFS Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP231993
Description
EFS Polyclonal specifically detects EFS in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EFS | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
O43281 | |
EFS | |
This antibody was developed against a recombinant protein corresponding to amino acids: IYKIPRASGTQLAAPRDALEVYDVPPTALRVPSSGPYDCPASFSHPLTRVAPQPPGEDDAPYDVPLTPKPPAELEPDLEWEGGREPGPPIYAAPSNLKRASALLNLYEAPEELLADGEGGGTDEGIYDVPLLGPEAPPSPEPPGALAS | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
CAS3, CASS3Cas scaffolding protein family member 3, EFS1, EFS2, embryonal Fyn-associated substrate, HEFS, signal transduction protein (SH3 containing), SIN | |
Rabbit | |
Affinity Purified | |
RUO | |
10278 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction